siRNA Details
Gene Symbol: LOC105378948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF148950.2 | - | 1 | 601 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY238224.1 | - | 1 | 90 | Chr. 1 - 1065976 - 1066443 [-] on Build GRCh38 |
