siRNA Details
Gene Symbol: LOC105378948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF148950.2 | - | 1 | 330 |
Gene Symbol: RNF223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001287.1 | XP_016856776.1 | 2 | 902 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN359402.1 | - | 1 | 604 | Chr. 1 - 1066638 - 1067258 [-] on Build GRCh38 |
