siRNA Details
Gene Symbol: GLUD1P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111968.1 | 4, 5 | 259 | |||
GenBank mRNA | BX281697.1 | - | 4, 5 | 259 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR604707.1 | - | 3, 4 | 1286 | Chr. 10 - 46759053 - 46786811 [-] on Build GRCh38 |
