siRNA Details
Gene Symbol: CHCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317178.1 | NP_001304107.1 | 5 | 923 | ||
GenBank mRNA | AK294071.1 | - | 5 | 805 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR611779.1 | - | 1 | 13 | Chr. 7 - 132972911 - 132973704 [-] on Build GRCh38 |
