siRNA Details
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031723.3 | NP_001026893.1 | 8 | 5511 | ||
NM_001278310.1 | NP_001265239.1 | 7 | 5339 | |||
GenBank mRNA | AK095074.1 | - | 1 | 1595 |
