siRNA Details
Gene Symbol: SAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174156.1 | NP_001167627.1 | 6 | 3008 | ||
NM_144660.2 | NP_653261.1 | 5 | 3235 | |||
XM_005269541.4 | XP_005269598.1 | 6 | 3199 | |||
XM_011539311.1 | XP_011537613.1 | 6 | 3127 | |||
XM_011539312.2 | XP_011537614.1 | 6 | 3057 | |||
XM_017015738.1 | XP_016871227.1 | 6 | 3064 | |||
XM_017015739.1 | XP_016871228.1 | 6 | 3092 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK055455.1 | - | 40 | Chr. 14 - 67770973 - 67772857 [-] on Build GRCh38 |
