siRNA Details
Gene Symbol: FLJ46365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128232.1 | - | 1 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB099368.1 | - | 1 | 221 | Chr. 8 - 48590401 - 48591510 [+] on Build GRCh38 |
