siRNA Details
Gene Symbol: PTPRD-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110696.1 | 3 | 2118 | |||
GenBank mRNA | BC045688.1 | - | 3 | 2118 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PTPRD-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110696.1 | 3 | 2118 | |||
GenBank mRNA | BC045688.1 | - | 3 | 2118 |