siRNA Details
Gene Symbol: LOC284630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096384.1 | - | 1 | 1322 |
Gene Symbol: TTC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_016999990.1 | XP_016855479.1 | 9 | 4243 |
