siRNA Details
Gene Symbol: LINC00642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033986.1 | 4 | 421 | |||
GenBank mRNA | AK093266.1 | - | 3 | 1739 | ||
BC036259.1 | - | 3 | 1088 | |||
BX101323.1 | - | 4 | 421 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033986.1 | 4 | 421 | |||
GenBank mRNA | AK093266.1 | - | 3 | 1739 | ||
BC036259.1 | - | 3 | 1088 | |||
BX101323.1 | - | 4 | 421 |