siRNA Details
Gene Symbol: LACTB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038881.1 | 1 | 143 | |||
GenBank mRNA | AL833348.1 | - | 1 | 143 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX090672.1 | - | 1 | 116 | Chr. 8 - 70608604 - 70652712 [+] on Build GRCh38 |
