siRNA Details
Gene Symbol: LOC728573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123270.1 | - | 1 | 917 |
Gene Symbol: LOC107985384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028202.1 | XP_016883691.1 | 1 | 960 |
