siRNA Details
Gene Symbol: LINC00298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043361.1 | - | 1 | 22 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF338305.1 | - | 1 | 21 | Chr. 2 - 7923917 - 7979309 [-] on Build GRCh38 |
