siRNA Details
Gene Symbol: LOC284108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093715.1 | - | 1 | 332 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR616125.1 | - | 1 | 320 | Chr. 17 - 39165085 - 39167849 [+] on Build GRCh38 |
