siRNA Details
Gene Symbol: LINC00508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126452.2 | 4 | 1078 | |||
GenBank mRNA | BC032019.1 | - | 1 | 767 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126452.2 | 4 | 1078 | |||
GenBank mRNA | BC032019.1 | - | 1 | 767 |