siRNA Details
Gene Symbol: LOC101928885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130778.1 | 1 | 36 | |||
GenBank mRNA | AK128486.1 | - | 1 | 36 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB212090.1 | - | 1 | 36 | Chr. 5 - 68970692 - 69029927 [-] on Build GRCh38 |
