siRNA Details
Gene Symbol: APOBEC3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094570.1 | - | 1 | 1065 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC007954.1 | - | 1 | 1302 | Chr. 22 - 39018842 - 39020352 [+] on Build GRCh38 |
