siRNA Details
Gene Symbol: MCF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112732.2 | NP_001106203.2 | 30 | 6069 | ||
NM_001320815.1 | NP_001307744.1 | 31 | 6176 | |||
NM_001320816.1 | NP_001307745.1 | 30 | 6278 | |||
NM_024979.4 | NP_079255.4 | 30 | 6101 | |||
GenBank mRNA | AF086413.1 | - | 1 | 330 |
