siRNA Details
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL355686.1 | - | 2 | 614 | ||
BC054341.1 | - | 1 | 910 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DN912953.1 | - | 1 | 519 | Chr. 21 - 44800868 - 44801406 [-] on Build GRCh38 |
