siRNA Details
Gene Symbol: LOC647070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001442.1 | - | 2 | 317 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU128475.1 | - | 2 | 317 | Chr. 1 - 182837404 - 182839591 [-] on Build GRCh38 |
