siRNA Details
Gene Symbol: LINC01206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104146.1 | 8 | 6242 | |||
GenBank mRNA | BX641067.1 | - | 1 | 2972 | ||
CR749396.1 | - | 1 | 2162 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104146.1 | 8 | 6242 | |||
GenBank mRNA | BX641067.1 | - | 1 | 2972 | ||
CR749396.1 | - | 1 | 2162 |