siRNA Details
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266930.2 | XP_005266987.1 | 9 | 7966 | ||
XM_017010727.1 | XP_016866216.1 | 8 | 7820 | |||
XM_017010728.1 | XP_016866217.1 | 11 | 8126 | |||
GenBank mRNA | AK055220.1 | - | 1 | 2584 |
