siRNA Details
Gene Symbol: NEMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021760.1 | XP_016877249.1 | 32 | 4951 | ||
XM_017021761.1 | XP_016877250.1 | 27 | 4404 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NEMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021760.1 | XP_016877249.1 | 32 | 4951 | ||
XM_017021761.1 | XP_016877250.1 | 27 | 4404 |