siRNA Details
Gene Symbol: LOC100506274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038432.1 | 3 | 314 | |||
GenBank mRNA | AK093290.1 | - | 3 | 311 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB029885.1 | - | 3 | 313 | Chr. 2 - 7421265 - 7428777 [+] on Build GRCh38 |
