siRNA Details
Gene Symbol: LOC101927410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110777.1 | 1 | 235 | |||
GenBank mRNA | AK056572.1 | - | 1 | 235 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA744757.1 | - | 1 | 236 | Chr. 18 - 10661933 - 10663885 [+] on Build GRCh38 |
