siRNA Details
Gene Symbol: LINC01116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040001.2 | 3 | 531 | |||
GenBank mRNA | BC013438.1 | - | 3 | 888 | ||
BC013588.1 | - | 1 | 200 | |||
BE908176.1 | - | 3 | 505 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040001.2 | 3 | 531 | |||
GenBank mRNA | BC013438.1 | - | 3 | 888 | ||
BC013588.1 | - | 1 | 200 | |||
BE908176.1 | - | 3 | 505 |