siRNA Details
Gene Symbol: LINC01116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040001.2 | 3 | 963 | |||
GenBank mRNA | BC013588.1 | - | 1 | 632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX113755.1 | - | 1 | 512 | Chr. 2 - 176629589 - 176630174 [-] on Build GRCh38 |
