siRNA Details
Gene Symbol: LINC01116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040001.2 | 3 | 561 | |||
GenBank mRNA | BC013438.1 | - | 3 | 918 | ||
BC013588.1 | - | 1 | 230 | |||
BE908176.1 | - | 3 | 535 | |||
BF029957.1 | - | 3 | 560 |
