siRNA Details
Gene Symbol: ZZEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001683.1 | - | 15 | 2614 | ||
BC047285.1 | - | 15 | 2777 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U33270.1 | - | 1 | 23 | Chr. 17 - 4021952 - 4022483 [-] on Build GRCh38 |
