siRNA Details
Gene Symbol: LOC101928009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110685.1 | 2 | 605 | |||
GenBank mRNA | BC038426.1 | - | 2 | 605 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG720230.1 | - | 2 | 610 | Chr. 1 - 153174518 - 153190097 [+] on Build GRCh38 |
