siRNA Details
Gene Symbol: LOC101928865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120564.1 | 2, 3 | 352 | |||
GenBank mRNA | BC039514.1 | - | 2, 3 | 352 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM454724.1 | - | 352 | Chr. - - [-] on Build |
