siRNA Details
Gene Symbol: LOC284933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038917.1 | 2 | 206 | |||
GenBank mRNA | BC036359.1 | - | 2 | 206 | ||
BC048428.1 | - | 2 | 138 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI830326.1 | - | 2 | 206 | Chr. 22 - 48544684 - 48547387 [-] on Build GRCh38 |
