siRNA Details
Gene Symbol: LOC101929199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125921.1 | 2 | 171 | |||
GenBank mRNA | AK125145.1 | - | 2 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA472556.1 | - | 2 | 172 | Chr. 4 - 27217479 - 27277054 [+] on Build GRCh38 |
