siRNA Details
Gene Symbol: MYHAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125367.1 | 6 | 475 | |||
GenBank mRNA | AK097500.1 | - | 6 | 463 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB033382.1 | - | 6 | 463 | Chr. 17 - 10383144 - 10598222 [+] on Build GRCh38 |
