siRNA Details
Gene Symbol: LOC102724659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126000.1 | 1 | 296 | |||
GenBank mRNA | BC034298.1 | - | 1 | 296 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI826945.1 | - | 1 | 296 | Chr. 1 - 11778219 - 11779619 [-] on Build GRCh38 |
