siRNA Details
Gene Symbol: RPS6P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052613.1 | - | 1 | 72 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ711654.1 | - | 1 | 85 | Chr. 4 - 82495363 - 82496279 [-] on Build GRCh38 |
