siRNA Details
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AJ412030.1 | - | LIT1922 BCMS |
3 | 511 | |
AJ412032.1 | - | LIT1916 BCMS |
4 | 686 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AJ411997.3 | - | 1 | 158 | Chr. 13 - 50408172 - 50408463 [+] on Build GRCh38 |
