siRNA Details
Gene Symbol: LOC105375092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056771.1 | - | 1 | 254 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA837430.1 | - | 1 | 255 | Chr. 6 - 53065824 - 53066385 [-] on Build GRCh38 |
