siRNA Details
Gene Symbol: LOC101927027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110204.1 | 5 | 2052 | |||
GenBank mRNA | CD676753.1 | - | 1 | 214 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK128542.1 | - | 1 | 4554 | Chr. 2 - 178435401 - 178440059 [+] on Build GRCh38 |
