siRNA Details
Gene Symbol: KDM4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF085959.1 | - | 1 | 312 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX115579.1 | - | 1 | 312 | Chr. 9 - 6908974 - 6909628 [+] on Build GRCh38 |
