siRNA Details
Gene Symbol: LOC101928524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055017.1 | - | 1 | 253 | ||
BX647538.1 | - | 1 | 3912 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA060916.1 | - | 1 | 253 | Chr. 10 - 13642305 - 13642854 [+] on Build GRCh38 |
