siRNA Details
Gene Symbol: LOC101928940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120567.1 | 2 | 242 | |||
GenBank mRNA | BC033908.1 | - | 2 | 247 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG718298.1 | - | 2 | 246 | Chr. 11 - 114363796 - 114380026 [-] on Build GRCh38 |
