siRNA Details
Gene Symbol: ZBTB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 24169 | ||
NM_001164343.2 | NP_001157815.1 | 12 | 24631 | |||
NM_001164344.2 | NP_001157816.1 | 8 | 24595 | |||
NM_001164345.2 | NP_001157817.1 | 7 | 24549 | |||
NM_001164346.2 | NP_001157818.1 | 6 | 24188 | |||
NM_001164347.2 | NP_001157819.1 | 6 | 24210 | |||
NM_015642.5 | NP_056457.3 | 10 | 24393 | |||
NR_121662.1 | 7 | 22651 | ||||
GenBank mRNA | BC016962.1 | - | 1 | 566 |
