siRNA Details
Gene Symbol: LOC255654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040002.1 | 3 | 311 | |||
GenBank mRNA | BC039539.1 | - | 3 | 311 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM908092.1 | - | 3 | 310 | Chr. 1 - 246516042 - 246524286 [-] on Build GRCh38 |
