siRNA Details
Gene Symbol: LINC01043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135321.1 | 3336 | ||||
GenBank mRNA | BX649107.1 | - | 3336 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135321.1 | 3336 | ||||
GenBank mRNA | BX649107.1 | - | 3336 |