siRNA Details
Gene Symbol: LOC101928043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110692.1 | 2 | 129 | |||
GenBank mRNA | BC039533.1 | - | 2 | 129 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM563376.1 | - | 123 | Chr. - - [-] on Build |
