siRNA Details
Gene Symbol: ARHGAP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256025.2 | NP_001242954.1 | 1 | 134 | ||
NR_045675.1 | 2 | 771 | ||||
XM_011540002.1 | XP_011538304.1 | 2 | 771 | |||
XM_011540003.1 | XP_011538305.1 | 2 | 73 | |||
XM_011540004.1 | XP_011538306.1 | 2 | 771 | |||
XM_011540006.1 | XP_011538308.1 | 2 | 771 | |||
XM_011540013.2 | XP_011538315.1 | 2 | 771 | |||
XM_011540015.2 | XP_011538317.1 | 2 | 771 | |||
XM_017016473.1 | XP_016871962.1 | 2 | 771 | |||
GenBank mRNA | AK055937.1 | - | 2 | 771 | ||
AK293579.1 | - | 1 | 20 | |||
DC391765.1 | - | 1 | 134 |
