siRNA Details
Gene Symbol: TIAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126861.1 | - | 1 | 573 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124161.1 | - | 1 | 2123 | Chr. 17 - 29076584 - 29078857 [-] on Build GRCh38 |
