siRNA Details
Gene Symbol: FAM199X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207318.3 | NP_997201.1 | 6 | 6949 | ||
XM_005262079.2 | XP_005262136.1 | 6 | 6817 | |||
XM_017029270.1 | XP_016884759.1 | 6 | 6518 | |||
GenBank mRNA | AB075510.1 | - | 2 | 1650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR611073.1 | - | 1 | 1203 | Chr. X - 104194047 - 104195599 [+] on Build GRCh38 |
