siRNA Details
Gene Symbol: FAM199X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207318.3 | NP_997201.1 | 6 | 5878 | ||
XM_005262079.2 | XP_005262136.1 | 6 | 5746 | |||
XM_017029270.1 | XP_016884759.1 | 6 | 5447 | |||
GenBank mRNA | AK024844.1 | - | 1 | 932 |
